Oxoid™ Skirrow Campylobacter Selective Supplement
Thermo Scientific™

Oxoid™ Skirrow Campylobacter Selective Supplement

Oxoid Campylobacter Selective Supplement, Skirrow prepares Skirrow Selective Medium for isolation of Campylobacter spp. at 42°C.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수율
SR0069HEach vial for 2L medium
SR0069EEach vial for 500mL medium
카탈로그 번호 SR0069H
제품 가격(KRW)
-
수율:
Each vial for 2L medium

Oxoid Campylobacter Selective Supplement, Skirrow is used for preparation of Skirrow Selective Medium for the isolation of Campylobacter spp. at 42°C.

Add to Blood Agar Base No. 2, Part No. CM0271B or Columbia Blood Agar Base, Part No. CM0331B
사양
설명Campylobacter Selective Supplement, Skirrow
수량10/Pk.
수율Each vial for 2L medium
형태Freeze-dried
제품 유형Selective Supplement
Unit SizeEach