Oxoid™ Perfringens OPSP Selective Supplement B
Thermo Scientific™
Oxoid™ Perfringens OPSP Selective Supplement B
Oxoid Perfringens OPSP Selective Supplement B is for selective isolation of Clostridium perfringens from food.
Have Questions?
Número do catálogoQuantity
SR0077E10/Pk.
Número do catálogo SR0077E
Preço (BRL)
-
Quantity:
10/Pk.
Oxoid Perfringens OPSP Selective Supplement B is for selective isolation of Clostridium perfringens from food.
  • Add to Perfringens Agar Base, OPSP, Part No. CM0543B
  • For 500mL of medium
For Laboratory Use
For Laboratory Use
Especificações
DescriptionPerfringens OPSP Supplement B
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesPerfringens
Unit SizeEach