Oxoid™ Chloramphenicol Selective Supplement
Actual product may vary
Thermo Scientific™
Oxoid™ Chloramphenicol Selective Supplement
Oxoid Chloramphenicol Selective Supplement assists in the isolation of yeast and molds associated with spoilage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberYield
SR0078EEach vial for 500mL medium
SR0078HEach vial for 2L medium
Catalog number SR0078E
Price (USD)
-
Yield:
Each vial for 500mL medium
For the isolation of yeast and molds associated with spoilage, add Oxoid Chloramphenicol Selective Supplement to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.
For the isolation of yeast and molds associated with spoilage, add to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.
Specifications
DescriptionChloramphenicol Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
Unit SizeEach