Oxoid™ Butzler Campylobacter Selective Supplement
Thermo Scientific™

Oxoid™ Butzler Campylobacter Selective Supplement

The Oxoid Campylobacter Selective Supplement, Butzler is used for the isolation of Campylobacter spp. at 37°C.
Have Questions?
Catalog NumberQuantity
SR0085E10/Pk.
Catalog number SR0085E
Price (CLP)
-
Quantity:
10/Pk.

Oxoid Campylobacter Selective Supplement, Butzler is used for the isolation of Campylobacter spp. at 37°C.

  • Add to Columbia Blood Agar Base, Part No. CM0331B
  • Each vial supplements 500mL of medium
Specifications
DescriptionCampylobacter Selective Supplement, Butzler
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesCampylobacter
Unit SizeEach