Oxoid™ Preston Campylobacter Selective Supplement
Thermo Scientific™
Oxoid™ Preston Campylobacter Selective Supplement
Used for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian, and environmental samples.
Have Questions?
Catalog NumberQuantity
SR0117E10/Pk.
Catalog number SR0117E
Price (MXN)
-
Quantity:
10/Pk.
Oxoid Preston Campylobacter Selective Supplement is used for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian, and environmental samples.
  • Add to Campylobacter Agar Base, Part No. CM0689B or Nutrient Broth No. 2, Part No. CM0067B
  • Each vial supplements 500mL of medium
Specifications
DescriptionPreston Campylobacter Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesCampylobacter
Unit SizeEach