Oxoid™ Modified Bolton Broth Selective Supplement
Thermo Scientific™
Oxoid™ Modified Bolton Broth Selective Supplement
Oxoid Modified Bolton Broth Selective Supplement is for the selective pre-enrichment of Campylobacter spp. in food samples.
Have Questions?
Catalog NumberQuantity
SR0208E10/Pk.
Catalog number SR0208E
Price (CLP)
-
Quantity:
10/Pk.
Oxoid Modified Bolton Broth Selective Supplement is for the selective pre-enrichment of Campylobacter spp. in food samples.
  • Alternative supplement containing amphotericin B as a replacement for cycloheximide
  • Add to Bolton Broth, Part No. CM0983B
  • Each vial supplements 500mL of medium
For Laboratory Use
For Laboratory Use
Specifications
DescriptionModified Bolton Broth Selective Supplement
Quantity10/Pk.
YieldEach vial for 500mL medium
FormFreeze-dried
Product TypeSelective Supplement
SpeciesBolton
Unit SizeEach