Oxoid Novobiocin Selective Supplement, Liquid, 10 mg
Thermo Scientific™
Oxoid Novobiocin Selective Supplement, Liquid, 10 mg
Selective supplement for variety of culture media.
Have Questions?
Número de catálogo SR0249A
Precio (USD)
-
Oxoid Novobiocin Selective Supplement (10mg/vial) can be added to a range of culture media such as:
BPW (ISO formulation) Part number CM1211 as part of the Salmonella Precis method EC Broth, Part No. CM0990B; Muller-Kauffmann Tetrathionate-Novobiocin Broth Base, Part No. CM0343B; Brilliant Green Agar, Part No. CM0263B; MRSV, Part No. CM1112B; Modified Tryptone Soya Broth, Part No. CM0989B; Lysine Iron Agar, Modified, Part No. CM0886B
Each vial contains 40 mL of liquid novobiocin that has a concentration of 10 mg.
Especificaciones
Cantidad10 Vials
FormularioLíquido
Tipo de productoMicrobiology Media
Unit SizeEach