Search
Search

Enable the performance of Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) with these selective and differential supplements.
| Número do catálogo | Yield | Form |
|---|---|---|
| SR0258E | 1 vial / 500 mL agar | Liquid |
| SR0257E | 1 vial / 500 mL agar | Pellets |
| SR0257B | 1 vial/ 200 mL BO1370Z medium | Pellets |
| SR0258B | 1 vial/ 200 mL BO1370Z medium | Liquid |
Enable the performance of Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Base with the Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Supplements.
When used to supplement the Brilliance™ Listeria Agar (ISO) Supplements, the Selective Supplement enables the inhibition of competing microflora whilst the Differential Supplement aids the differentiation of Listeria monocytogenes from other Listeria species by facilitating the development of an opaque white halo around the colonies.
Enable the performance of the Thermo Scientific™ Oxoid™ Brilliance™ Agar (ISO) Base with the Brilliance™ Listeria Agar (ISO) Supplements.