Speedvac™ SRF110 Kühl-Zentrifugalvakuumkonzentrator und Rotoren
Speedvac™ SRF110 Kühl-Zentrifugalvakuumkonzentrator und Rotoren
Speedvac™ SRF110 Kühl-Zentrifugalvakuumkonzentrator und Rotoren
Thermo Scientific™

Speedvac™ SRF110 Kühl-Zentrifugalvakuumkonzentrator und Rotoren

Verarbeiten Sie hitzeempfindliche biologische Proben zuverlässig mit dem SRF110. Dieses gekühlte SpeedVac-System nutzt modernste Zentrifugations-, Vakuum- und Wärmetechnologien, um Lösungsmittel zu entfernen und wärmeempfindliche biologische Proben zu konzentrieren, während die Probenintegrität erhalten bleibt.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerTypElektrische AnforderungenEnthält
SRF110P2-230Universal-Vakuumkonzentrator-Kit230 V 50 Hz 8 AUniversal-Vakuumsystem, Rotor Zubehör
RF10812-13 mm RotorNicht zutreffendRotor
RF132Hexagonaler MikrozentrifugenrotorNicht zutreffendRotor
RF3228 mm RotorNicht zutreffendRotor
RF5212-17 mm RotorNicht zutreffendRotor
RFMT4MikrotiterplattenrotorNicht zutreffendRotor
SRF110-115Vakuumkonzentrator115 V, 60 Hz, 16 AKonzentrator
SRF110-230Vakuumkonzentrator230 V 50 Hz 8 AKonzentrator
SRF110P1-115Vakuumkonzentrator-Kit115 V, 60 Hz, 16 AÖlpumpe, Dampfabscheider, Zubehör
SRF110P1-230Vakuumkonzentrator-Kit230 V 50 Hz 8 AÖlpumpe, Dampfabscheider, Zubehör
SRF110P2-115Universal-Vakuumkonzentrator-Kit115 V, 60 Hz, 16 AUniversal-Vakuumsystem, Rotor Zubehör
Katalognummer SRF110P2-230
Preis (EUR)
-
Typ:
Universal-Vakuumkonzentrator-Kit
Elektrische Anforderungen:
230 V 50 Hz 8 A
Enthält:
Universal-Vakuumsystem, Rotor Zubehör

SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.

SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space.  SRF110 can be configured depending on your applications need. Select from wide range of suitable Thermo Scientific components such as pumps, Vapor Traps, and additional accessories to configure your custom vacuum concentration setup.  For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.

  • Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
  • 3 Preset program with option to store up to 9 programs
  • Temperature range -4°C up to 100°C (1°C increment)
  • Self aligning and quick release rotor mechanism
  • Hexagonal rotor (RF132) included with purchase of SRF110
  • Refer to list of rotors available for purchase and to be used with SRF110 model
  • Electricals- 115V AC (60Hz, 16Amps) and 230V AC (50Hz, 8Amps)
  • Certifications- UL Certified, CE, UKCA, KC, RCM Marked.
  • F Gas Compliance- Yes (EPA-SNAP and EU)
  • SRF110P1 kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), an Oil pump (VLP120-115), a vapor Trap (RVT450-115), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
  • SRF110P2 kit includes SRF110P2 Kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), Universal Vacuum System (UVS450), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
Specifications
ZertifizierungenCE, UKCA, KC, RCM
BeschreibungKit bestehend aus einem gekühlten Zentrifugalvakuumkonzentrator mit mittlerer Kapazität, Universalvakuumsystem, Rotor, Zubehör, 230 V, 50 Hz, 8 A
Abmessungen (L x B x H)56 x 37 x 43 cm
Elektrische Anforderungen230 V 50 Hz 8 A
EnthältUniversal-Vakuumsystem, Rotor Zubehör
MaterialPulverbeschichteter Stahl
Versandgewicht (angloamerikanisch)108 lb.
Versandgewicht (metrisch)49 kg
TypUniversal-Vakuumkonzentrator-Kit
Gewicht (angloamerikanisch)95 lb.
Gewicht (metrisch)43 kg
KammermaterialBeschichtete Aluminiumkammer
Temperaturbereich (metrisch)-4 °C bis +100 °C
Unit SizeEach