Please note: order placement quantity must be in multiples of 10, minimum of 10 required.
2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, ReadyMate connectors and injection site
Thermo Scientific™

2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, ReadyMate connectors and injection site

The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SS00109I
also known as SS00109-I
50 L
SS00110I
also known as SS00110-I
100 L
SS00111I
also known as SS00111-I
200 L
Catalog number SS00109I
also known as SS00109-I
Price (USD)
-
Capacity (Metric):
50 L
The 50 L Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (2 ReadyMate pinch clamps, 1 injection site).

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: ReadyMate, pinch clamp. Tubing: C-Flex, length: 51 cm (20 in.), ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
• Line 2: ReadyMate, pinch clamp. Tubing: C-Flex, length: 51 cm (20 in.), ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.)
• Line 3: Tubing: C-Flex, length 7.6 cm (3 in.), 6.4 mm (1/4 in.) luer injection port

Specifications
Dimensions95.3 cm x 54.6 cm (37.5 in. x 21.5 in.)
For Use With (Equipment)TK-0050-05
Outer Packaging4 mil. poly bag, One 4-mil. poly bag
Product LineLabtainer
Protective Inner PackagingPoly bags on tubing ends, protective material around rough areas
Quantity10 per case
Shipping ConditionRoom Temperature
Sterilization MethodIrradiated
Capacity (Metric)50 L
Film DescriptionASI 26/77
MaterialPolyethylene
Port DetailReadyMate DAC connections and injection port
Tubing StyleC-Flex, 51 cm (20 in.)
Unit Size50 L
Contents & Storage
Store at room temperature.