Orion Star T900 Series Titrator Accessories
Actual product may vary
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Orion Star T900 Series Titrator Accessories
Thermo Scientific™

Orion Star T900 Series Titrator Accessories

Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
START-PS4ANZ Power Supply
START-B00Burette Cover
START-B1010 mL Burette
START-B2020 mL Burette
START-B5050 mL Burette
START-BT0Reagent Bottle Holder
START-BT11 L Plastic Bottle
START-BT21 L Glass Bottle
START-BT312 x 1 L Plastic Bottles
START-BT412 x 1 L Glass Bottles
START-CP1Reagent Bottle Cap (GL38)
START-CP2Reagent Bottle Cap (GL45)
START-CP3Pint Bottle Cap
START-DS1Dispenser Probe
START-DVKDispenser Verification Kit
START-EH1Electrode Holder Assembly
START-EH2Stopper
START-EH3Cable Management
START-PS1US/Japan Power Supply
START-PS2EU Power Supply
START-PS3UK/Singapore Power Supply
START-PS5China Power Supply
START-TB1Standard Tubing Kit
START-TB2Light-blocking Tubing Kit
START-TB3Drying Tube
START-TB4Replacemet Burette
START-TB5Replacement Reagent Cap
START-TB6Replacement Dispenser Probe
START-UM1User Manual
START-USBUSB Cable
START-VK1Value Replacement
Catalog number START-PS4
Price (USD)
42.75
Online Exclusive
47.50
Save 4.75 (10%)
Each
Type:
ANZ Power Supply
Price (USD)
42.75
Online Exclusive
47.50
Save 4.75 (10%)
Each
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
  • Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
  • Bottle holder helps secure titrant to prevent spills and offers flexible placement
  • Color-coded tubing and connectors simplify tubing installation and maintenance
Specifications
DescriptionOrion Star T900 series 230V Australia/New Zealand power supply
For Use With (Equipment)Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator
TypeANZ Power Supply
Unit SizeEach