SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, PVDF
Invitrogen™

SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.2 μm, PVDF

Invitrogen SureLock Tandem Midi Membranes and Filters, 0.2 μm, PVDF, have excellent binding properties for western blotting, dot-blot assays, andRead more
Have Questions?
Catalog NumberDimensions (LxW)
STM2005Midi, 14 cm x 8.6 cm
Catalog number STM2005
Price (USD)
344.00
1 each
Dimensions (LxW):
Midi, 14 cm x 8.6 cm
Price (USD)
344.00
1 each
Invitrogen SureLock Tandem Midi Membranes and Filters, 0.2 μm, PVDF, have excellent binding properties for western blotting, dot-blot assays, and protein sequencing. The 0.2 μm pore size makes these membranes ideal for protein analysis of small amounts of protein (as little as 10 pmoles), amino acid analysis, and transfer of small molecular weight proteins and peptides. The dimensions (8.6 x 14 cm) of these PVDF membrane and filter paper sandwiches are ideal for wet-transfer of Invitrogen midi gels using the SureLock Tandem Midi Blot Module. The 0.2 μm pore size membrane and filter paper sandwiches are conveniently supplied as pack of 20.

Features
• Membrane: polyvinylidene difluoride (PVDF)
• Filter paper thickness: 0.8 mm
• Re-probe characteristics: yes
• Pre-activation: required with 100% alcohol (methanol, ethanol, or isopropanol)
• Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• Durability: compatible with most organic solvents, acids, and mild bases
• Dimensions: 8.6 x 14 cm
Specifications
Quantity20 Pre-cut Membranes and Filters, 0.2 μm, PVDF Sandwich
Dimensions (LxW)Midi, 14 cm x 8.6 cm
FormatSandwich
MaterialPVDF
Pore Size0.2 μm
Product LineSureLock
Thickness0.8 mm
Unit Size1 each
Contents & Storage
Contains 20 pre-cut membrane and filter sandwiches (0.2 μm, PVDF)