SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, PVDF
Invitrogen™

SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, PVDF

Invitrogen SureLock Tandem Midi Membranes and Filters, 0.45 μm, PVDF, have excellent binding properties for western blotting, dot-blot assays, andRead more
Have Questions?
Catalog NumberDimensions (LxW)
STM2006Midi, 14 cm x 8.6 cm
Catalog number STM2006
Price (USD)
344.00
1 each
Dimensions (LxW):
Midi, 14 cm x 8.6 cm
Price (USD)
344.00
1 each
Invitrogen SureLock Tandem Midi Membranes and Filters, 0.45 μm, PVDF, have excellent binding properties for western blotting, dot-blot assays, and protein sequencing. The 0.45-μm pore size makes these membranes ideal for protein transfer for proteins >10 kDa. The dimensions (8.6 x 14 cm) of these PVDF membrane and filter paper sandwiches are ideal for wet-transfer of Invitrogen midi gels using the SureLock Tandem Midi Blot Module. The 0.45-μm pore size membrane and filter paper sandwiches are conveniently supplied as pack of 20.

Features
• Membrane: polyvinylidene difluoride (PVDF)
• Filter paper thickness: 0.8 mm
• Re-probe characteristics: yes
• Pre-activation: required with 100% alcohol (methanol, ethanol, or isopropanol)
• Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• Durability: compatible with most organic solvents, acids, and mild bases
• Dimensions: 8.6 x 14 cm
Specifications
Quantity20 Pre-cut Membranes and Filters, 0.45 μm, PVDF Sandwich
Dimensions (LxW)Midi, 14 cm x 8.6 cm
FormatSandwich
MaterialPVDF
Pore Size0.45 μm
Product LineSureLock
Unit Size1 each
Contents & Storage
Contains 20 pre-cut membrane and filter sandwiches (0.45 μm, PVDF)

Frequently asked questions (FAQs)

Do you offer pre-cut membranes and filters for the SureLock Tandem Midi Blot Module?

We offer the following pre-cut membranes and filters for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, nitrocellulose, Cat. No. STM2008
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 µm, nitrocellulose, Cat. No. STM2007
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, PVDF, Cat. No. STM2006

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.