SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, nitrocellulose
Invitrogen™

SureLock™ Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, nitrocellulose

Invitrogen SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, nitrocellulose, are high-quality membranes ideal for blotting proteins and nucleicRead more
Have Questions?
Catalog NumberDimensions (LxW)
STM2008Midi, 14 cm x 8.6 cm
Catalog number STM2008
Price (USD)
344.00
1 each
Dimensions (LxW):
Midi, 14 cm x 8.6 cm
Price (USD)
344.00
1 each
Invitrogen SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 μm, nitrocellulose, are high-quality membranes ideal for blotting proteins and nucleic acids. The 0.45-μm pore size is ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp). The dimensions (8.6 x 14 cm) of these nitrocellulose membrane and filter paper sandwiches are ideal for wet-transfer of Invitrogen midi gels using the SureLock Tandem Midi Blot Module. The 0.2 μm pore size membrane and filter paper sandwiches are conveniently supplied as pack of 20.

Features:
• Membrane: 100% pure nitrocellulose
• Filter paper thickness: 0.8 mm
• Re-probe characteristics: yes
• Binding interactions: hydrophobic and electrostatic
• Compatibility: compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• Dimensions: 8.6 x 14 cm
Specifications
Quantity20 Pre-cut Membranes and Filters, 0.45 μm, Nitrocellulose Sandwich
Dimensions (LxW)Midi, 14 cm x 8.6 cm
FormatSandwich
MaterialNitrocellulose
Pore Size0.45 μm
Product LineSureLock
Unit Size1 each
Contents & Storage
Contains 20 pre-cut membrane and filter sandwiches (0.45 μm, nitrocellulose)

Frequently asked questions (FAQs)

Do you offer pre-cut membranes and filters for the SureLock Tandem Midi Blot Module?

We offer the following pre-cut membranes and filters for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, nitrocellulose, Cat. No. STM2008
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 µm, nitrocellulose, Cat. No. STM2007
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, PVDF, Cat. No. STM2006

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.