Thermo Scientific™ Enhanced Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 10-inch exhaust filters
Thermo Scientific™
Thermo Scientific™ Enhanced Single-Use Fermentor BioProcess Container, Aegis™ 5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 10-inch exhaust filters
This Thermo Scientific Enhanced Single-Use Fermentor (eS.U.F.) BioProcess Container (BPC) is composed of Aegis 5-14 film (a five-layer, animal-origin freeRead more
Have Questions?
Catalog NumberQuantity
SUT00008300 L
Catalog number SUT00008
Price (USD)
-
Quantity:
300 L
This Thermo Scientific Enhanced Single-Use Fermentor (eS.U.F.) BioProcess Container (BPC) is composed of Aegis 5-14 film (a five-layer, animal-origin free film) and includes a re-designed turbine impeller system, a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, two 10-inch high flow exhaust filters (one as back-up if using a pinch clamp), and a clear 4-mm optical density window and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom that complies with cGMP standards as per ISO13485.
Features of this eS.U.F. BPC include:
• Utilizes three enhanced turbine impellers with power efficient design
• Provides oxygen delivery as good or better than stainless steel fermenters
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and gas filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Capacity (Metric)300 L
For Use With (Application)Fermentation
Latex FreeYes
Quantity300 L
AutoclavableNot Autoclavable
ColorClear
DisposableSingle-use
MaterialAegis 5-14
Product TypeBioprocess Container for Liquid
SterilitySterile
Unit Size300 L