Product Image
Thermo Scientific™

HyPerforma™ Drum Accessories, HyPerforma Support Plate for Conical HDPE Drums; For use with 200L Drum

The Thermo Scientific™ HyPerforma Support Plate for 200L Conical HDPE Drums enables easy transport of BioProcess Container Systems™ (BPC™s).Features:• RoundRead more
Have Questions?
Catalog NumberQuantity
SV5010202
also known as SV50102.02
200 L
Catalog number SV5010202
also known as SV50102.02
Price (USD)
-
Quantity:
200 L
The Thermo Scientific™ HyPerforma Support Plate for 200L Conical HDPE Drums enables easy transport of BioProcess Container Systems™ (BPC™s).

Features:
• Round plastic disk for use with conical HDPE drums with 4 ″ port
• Supports up to 200L drums

Specifications
Capacity (Metric)50, 100, 200 L
Port Detail4 in.
Product LineHyPerforma
Quantity200 L
For Use With (Equipment)Drum, 200 L
Unit Size200 L