Smart-Vue Pro Cloud Software and Subscription Services
Thermo Scientific™
Smart-Vue Pro Cloud Software and Subscription Services
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
SVPSWCS20500Up to 500 sensors
SVPSWCS20005Up to 5 sensors
SVPSWCS20020Up to 20 sensors
SVPSWCS20050Up to 50 sensors
SVPSWCS20100Up to 100 sensors
SVPSWCS20250Up to 250 sensors
SVPSWCS21000Up to 1000 sensors
SVPSWCS21500Up to 1500 sensors
SVPSWCS22000Up to 2000 sensors
Catalog number SVPSWCS20500
Price (USD)
-
For Use With (Application):
Up to 500 sensors
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times.
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • Audit trail and secure-level access control to support 21CFR Part 11
  • Cloud-based configurations allow flexibility to check status anytime, anywhere from any modern web browser.
Specifications
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
DescriptionSmart-Vue Pro Cloud license key - Access to the Cloud and Smart-Vue Pro Web application for installation up to 500 sensors. License validity: 12 months after activation
For Use With (Application)Up to 500 sensors
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Unit SizeEach