Smart-Vue Pro On-Premise Software and Subscription Services
Thermo Scientific™
Smart-Vue Pro On-Premise Software and Subscription Services
A simplified software experience, allowing you to focus on what matters the most
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
SVPSWPS20050On-premise LicenseUp to 50 measurement points
SVPSWPS20005On-premise LicenseUp to 5 measurement points
SVPSWPS20020On-premise LicenseUp to 20 measurement points
SVPSWPS20100On-premise LicenseUp to 100 measurement points
SVPSWPS20250On-premise LicenseUp to 250 measurement points
SVPSWPS20500On-premise LicenseUp to 500 measurement points
SVPSWPS21000On-premise LicenseUp to 1000 measurement points
SVPSWPS21500On-premise LicenseUp to 1500 measurement points
SVPSWPS22000On-premise LicenseUp to 2000 measurement points
SVPSWPU20020On-premise License Upgrade5 to 20 measurement points
SVPSWPU20050On-premise License Upgrade20 to 50 measurement points
SVPSWPU20100On-premise License Upgrade50 to 100 measurement points
SVPSWPU20250On-premise License Upgrade100 to 250 measurement points
SVPSWPU20500On-premise License Upgrade250 to 500 measurement points
SVPSWPU21000On-premise License Upgrade500 to 1000 measurement points
SVPSWPU21500On-premise License Upgrade1000 to 1500 measurement points
SVPSWPU22000On-premise License Upgrade1500 to 2000 measurement points
SVPSWSM20005Maintenance ContractUp to 5 measurement points
SVPSWSM20020Maintenance ContractUp to 20 measurement points
SVPSWSM20050Maintenance ContractUp to 50 measurement points
SVPSWSM20100Maintenance ContractUp to 100 measurement points
SVPSWSM20250Maintenance ContractUp to 250 measurement points
SVPSWSM20500Maintenance ContractUp to 500 measurement points
SVPSWSM21000Maintenance ContractUp to 1000 measurement points
SVPSWSM21500Maintenance ContractUp to 1500 measurement points
SVPSWSM22000Maintenance ContractUp to 2000 measurement points
Catalog number SVPSWPS20050
Price (USD)
-
Type:
On-premise License
Includes:
Up to 50 measurement points
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions the system will automatically notify the user based on prescribed notification preferences and allows you to document notes around each specific event for audit purposes. The floorplan view and local audio visual alarms will make it easy to locate alarming equipment during the critical recovery times. The on-premise solution maintains your Smart-Vue Pro data locally on your site's server.
  • Intuitive dashboard simplifies daily usage.
  • Optional floor plan and sites views.
  • Data can be exported with a click for reporting and customizable graphs.
  • Web application on Cloud or customer server.
  • Instant access to alarms, readings, and graphs.
  • Remote data access.
  • Flexible organization by site, department user, equipment, and criticality.
  • Audit trail and secure-level access control to support 21CFR Part 11
Specifications
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
DescriptionSmart-Vue Pro Web application on-premise license up to 50 measurement points
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
IncludesUp to 50 measurement points
TypeOn-premise License
Unit SizeEach