Smart-Vue Pro Alert Software
Thermo Scientific™
Smart-Vue Pro Alert Software
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
SVPSWSA21000Up to 1000 PTS
SVPSWSA20005Up to 5 PTS
SVPSWSA20020Up to 20 PTS
SVPSWSA20050Up to 50 PTS
SVPSWSA20100Up to 100 PTS
SVPSWSA20250Up to 250 PTS
SVPSWSA20500Up to 500 PTS
SVPSWSA21500Up to 1500 PTS
SVPSWSA22000Up to 2500 PTS
Catalog number SVPSWSA21000
Price (USD)
-
For Use With (Application):
Up to 1000 PTS
The Smart-Vue Pro system enables you to focus on your task at hand and does the sample conditions monitoring for you. In a case of issues with the defined sample conditions, Smart-Vue™ Alert will automatically notify the user based notification preferences wherever you are (phone, text or email).
  • Add-on notification package through SMS and voice calls (Twillo)
  • 12 month license (post-activation)
  • License depends on the number of monitoring points desired
Specifications
Certifications/ComplianceSmart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
DescriptionSmart-Vue Alert Software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors; License validity: 12 months after activation; Up to 1000 PTS
For Use With (Application)Up to 1000 PTS
For Use With (Equipment)Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
TypeSmart-Vue Pro Software and Subscription Service
Unit SizeEach