Smart-Vue Pro Laboratory Remote Monitoring PRO Kits
Thermo Scientific™
Smart-Vue Pro Laboratory Remote Monitoring PRO Kits
Smart-Vue Pro laboratory remote monitoring system is ideal for regulated environments in need of 21 CFR Part 11 support with audit trail. The IT friendly LoRaWan system provides easy set-up for a reliable, independent and secure wireless network.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberData MemoryCertifications/ComplianceIncludes
SVPULT5EU4000 Readings/Sensor, 40,000 ReadingsCertified for EMEA OnlyETH/4G LoRa Receiver Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, No SIM/Data
SVPREFFRZKT5EU4000 Readings/Sensor, 40,000 ReadingsCertified for EMEA OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, No SIM/Data
SVPULTKT1EU4000 Readings/Sensor, 8000 ReadingsCertified for EMEA OnlyETH/4G LoRa Receiver Gateway Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, No SIM/Data
SVPREFFRZKT1EU4000 Readings/Sensor, 8000 ReadingsCertified for EMEA OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, No SIM/Data
SVPULT5EUC4000 Readings/Sensor, 40,000 ReadingsCertified for EMEA OnlyETH/4G LoRa Receiver Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPREFFRZ5EUC4000 Readings/Sensor, 40,000 ReadingsCertified for EMEA OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPULTKT1EUC4000 Readings/Sensor, 8000 ReadingsCertified for EMEA OnlyETH/4G LoRa Receiver Gateway Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPREFFRZKT1EUC4000 Readings/Sensor, 8000 ReadingsCertified for EMEA OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPULTKT5NA4000 Readings/Sensor, 40,000 ReadingsCertified for North America OnlyETH/4G LoRa Receiver Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, No SIM/Data
SVPREFFRZKT5NA4000 Readings/Sensor, 40,000 ReadingsCertified for North America OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, No SIM/Data
SVPULTKT1NA4000 Readings/Sensor, 8000 ReadingsCertified for North America OnlyETH/4G LoRa Receiver Gateway Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, No SIM/Data
SVPREFFRZKT1NA4000 Readings/Sensor, 8000 ReadingsCertified for North America OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, No SIM/Data
SVPULTKT5NAC4000 Readings/Sensor, 40,000 ReadingsCertified for North America OnlyETH/4G LoRa Receiver Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPREFFRZKT5NAC4000 Readings/Sensor, 40,000 ReadingsCertified for North America OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPULTKT1NAC4000 Readings/Sensor, 8000 ReadingsCertified for North America OnlyETH/4G LoRa Receiver Gateway Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
SVPREFFRZKT1NACN/ACertified for North America OnlyETH/4G LoRa Gateway Kit, Cloud and Alerts Subscription, Power Supply, Antenna, SIM/Data for up to 10 Devices/Yr.
Catalog number SVPULT5EU
Price (CLP)
-
Data Memory:
4000 Readings/Sensor, 40,000 Readings
Certifications/Compliance:
Certified for EMEA Only
Includes:
ETH/4G LoRa Receiver Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, No SIM/Data
Smart-Vue Pro laboratory remote monitoring system is ideal for regulated environments in need of 21 CFR Part 11 support with audit trail. The IT friendly LoRaWan system provides easy set-up for a reliable, independent and secure wireless network.
Hardware
Receivers
Multitech Advanced receivers (blue) support up to 1000 data logging sensors. Gateways are available in 915 MHz for North America and 865-868 MHz for rest of world.
Multitech Pro receivers (white) Support for up to 50 data logging sensors per gateway and are available in Ethernet and 4G options in 915 MHz for North America and 865-868 MHz for rest of world.
Modules
Smart-Vue Pro modules collect data from independent sensors connected to equipment from any OEM. Smart-Vue Pro modules are available in two configurations- Duo and Quatro. The Duo allows up to 2 sensor channels for monitoring. The Quatro has extended range of 4 sensor channels for 2 wired and 2 wireless sensors. Ideal for monitoring several pieces of equipment simultaneously or multiple points within one piece of equipment.
Sensors
The Smart-Vue Pro sensors collect data points to be logged from any laboratory equipment OEM. Sensor kits, including gateways are available.
Subscription
Requires subscription
Availability
Check with your specialist for country availability. Receivers are available in 865-868 MHz and 915 MHz.
Specifications
Battery TypeLithium Ion
CalibrationNA
Certifications/ComplianceCertified for EMEA Only
Data Memory4000 Readings/Sensor, 40,000 Readings
For Use With (Equipment)ULT with 5 SVP Duo
IP RatingIP66/68
IncludesETH/4G LoRa Receiver Kit, Cloud and Alerts Subscription, Calibrated Emerald Data Logger, Power Supply, Antenna, No SIM/Data
ModelPRO MTC Receiver for up to 50 Modules
Operating Frequency865-868 Mhz
Quantity1
Sensor TypesCalibrated PT100: -100/+150
Warranty1 Year Warranty
Alarm TypesE-mail, SMS, Audible, Telephone, Mobile Push Notification
Display TypeLCD
TypeRemote Monitoring Gateway Bundle
Unit SizeEach