Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200
Thermo Scientific™

Beveled Non-Filtered Pipette Tips, 1-200 μL, QSP 200

Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityFormat
T090-QYellowNon-sterileBagged
T070-QNaturalNon-sterileBagged
T070RLN-QNaturalNon-sterileRefill
T070RLNS-QNaturalSterileRefill
T070RNS-QClearSterileHinged Rack
T090R-QYellowNon-sterileHinged Rack
T090RL-QYellowNon-sterileRefill
T090RLS-QYellowSterileRefill
T090RS-QYellowSterileHinged Rack
T110BRLS-QYellowSterileRefill
T110BRS-QYellowSterileHinged Rack
Catalog number T090-Q
Price (MXN)
-
Color:
Yellow
Sterility:
Non-sterile
Format:
Bagged
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.

Featuring:

  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design

Clean Claims:

QSP Pipette Tips are free of:

  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP

To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.

Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.

Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 200, Non-Filtered
GraduatedYes
Length (Metric)5.1 cm
ColorYellow
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatBagged
MaterialVirgin Polypropylene
Product LineQSP
SterilityNon-sterile
Tip StyleBeveled
Volume (Metric)200 μL
Unit SizeCase of 10000