Alpha-Bungarotoxin Conjugates
Invitrogen™

Alpha-Bungarotoxin Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or Dye
T1175Tetramethylrhodamine
B56130Alexa Fluor™ Plus 405
B13422Alexa Fluor™ 488
B35451Alexa Fluor™ 555
B13423Alexa Fluor™ 594
B35450Alexa Fluor™ 647
B1196Biotin-XX
B1601Unlabeled
Catalog number T1175
Price (USD)
513.32
Special offer
Online exclusive
Ends: 31-Dec-2026
626.00
Save 112.68
500 μg
Label or Dye:
Tetramethylrhodamine
Price (USD)
513.32
Special offer
Online exclusive
Ends: 31-Dec-2026
626.00
Save 112.68
500 μg
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.

Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor. Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorYellow
Detection MethodFluorescence
For Use With (Equipment)Fluorescence Microscope
Label or DyeTetramethylrhodamine
Product TypeTetramethylrhodamine α-Bungarotoxin
Quantity500 μg
Shipping ConditionRoom Temperature
Sub Cellular LocalizationReceptors
Excitation/Emission554/577 nm
Unit Size500 μg
Contents & Storage
Store in freezer (–5° to –30°C) and protect from light.

Citations & References (177)

Citations & References
Abstract
Acetylcholine receptor aggregation at nerve-muscle contacts in mammalian cultures: induction by ventral spinal cord neurons is specific to axons.
Authors:Dutton EK, Uhm CS, Samuelsson SJ, Schaffner AE, Fitzgerald SC, Daniels MP
Journal:J Neurosci
PubMed ID:7472493
We used a novel mammalian coculture system to study ACh receptor (AChR) redistribution and synaptic structure at nerve-muscle contacts. Ventral spinal cord (VSC) neurons were plated on cultures containing extensive myotubes but few fibroblasts. Neurite-induced redistribution of AChRs occurred within 6 hr after plating neurons and was maximal between 36-48 ... More
Signaling by insulin-like growth factors in paralyzed skeletal muscle: rapid induction of IGF1 expression in muscle fibers and prevention of interstitial cell proliferation by IGF-BP5 and IGF-BP4.
Authors:Caroni P, Schneider C
Journal:J Neurosci
PubMed ID:7514217
'In the absence of muscle activity, muscle fibers, muscle interstitial cells, and intramuscular nerves display characteristic reactions presumably aimed at restoring a functioning neuromuscular system and avoiding degenerative events. In partially denervated muscle these include proliferation of interstitial cells, followed by nerve sprouting. The same reactions can be induced in ... More
Identification of nicotinic acetylcholine receptor recycling and its role in maintaining receptor density at the neuromuscular junction in vivo.
Authors:Bruneau E, Sutter D, Hume RI, Akaaboune M
Journal:J Neurosci
PubMed ID:16251443
'In the CNS, receptor recycling is critical for synaptic plasticity; however, the recycling of receptors has never been observed at peripheral synapses. Using a novel imaging technique, we show here that nicotinic acetylcholine receptors (AChRs) recycle into the postsynaptic membrane of the neuromuscular junction. By sequentially labeling AChRs with biotin-bungarotoxin ... More
The dystroglycan complex is necessary for stabilization of acetylcholine receptor clusters at neuromuscular junctions and formation of the synaptic basement membrane.
Authors:Jacobson C, Côté PD, Rossi SG, Rotundo RL, Carbonetto S
Journal:J Cell Biol
PubMed ID:11157973
'The dystrophin-associated protein (DAP) complex spans the sarcolemmal membrane linking the cytoskeleton to the basement membrane surrounding each myofiber. Defects in the DAP complex have been linked previously to a variety of muscular dystrophies. Other evidence points to a role for the DAP complex in formation of nerve-muscle synapses. We ... More
Long-term maintenance of presynaptic function in the absence of target muscle fibers.
Authors:Dunaevsky A, Connor EA
Journal:J Neurosci
PubMed ID:7666196
'Here we have investigated the role of the muscle fiber in the maintenance of presynaptic function at frog motor nerve terminals. Muscle fibers were selectively damaged and prevented from regenerating while leaving the motor innervation intact. Presynaptic activity of the resulting target-deprived nerve terminals was assayed using the fluorescent dye, ... More