Tubulin Tracker™ Deep Red, 60 slides - FAQs
Tubulin Tracker™ Deep Red, 60 slides - FAQs
View additional product information for Tubulin Tracker™ Deep Red - FAQs (T34076, T34077)
4 product FAQs found
With Tubulin Tracker Deep Red and Tubulin Tracker Green, can I fix samples after labeling?
No. The labeling is not retained upon fixation. Imaging of the samples should be done only on live cells. To limit photobleaching while viewing live cells, use ProLong Live Antifade Reagent, for live cell imaging (Cat No. P36974 and P36975).
How is Docetaxel in Tubulin Tracker Deep Red different from Taxol in Tubulin Tracker Green? Are there any differences in the binding or cytotoxicity between the two reagents?
Docetaxel has a slightly higher binding affinity than Taxol and exhibits longer retention time due to better uptake and slower efflux, but for use in labeling cultured cells, these differences should be minimal. More information comparing the properties of these two reagents is provided in this publication: https://www.ncbi.nlm.nih.gov/pubmed/1671606 https://www.cell.com/fulltext/S1074-5521(05)00301-7
Is Tubulin Tracker Deep Red acetylated? Does it require intracellular esterase activity to make the reagent active?
Tubulin Tracker Deep Red is not acetylated. It is inherently cell permeable and is not modified further by any cellular enzymatic activity to fluoresce or bind to actin.
Do I need to use Probenecid when using Tubulin Tracker Deep Red?
The use of Probenecid is optional. Its requirement is dependent upon the cell type. Some cell types actively efflux the reagent out of the cells within a short time, limiting detection, other cell types do not exhibit this activity or exhibit very limited activity.