What is the concentration of TNP-ATP (2'-(or-3')-O-(Trinitrophenyl) Adenosine 5'-Triphosphate, Trisodium Salt) (Cat. No. T7602)?
The concentration is 5 mg/mL. This information can also be found on the product COA.