Dionex™ AS-AP Autosampler eLearning Courses
Dionex™ AS-AP Autosampler eLearning Courses
Dionex™ AS-AP Autosampler eLearning Courses
Dionex™ AS-AP Autosampler eLearning Courses
Thermo Scientific™

Dionex™ AS-AP Autosampler eLearning Courses

The Dionex™ AS-AP autosampler eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TASAMTSUser Maintenance and Troubleshooting
TASAPBOPBasic Operations
Catalog number TASAMTS
Price (USD)
130.00
Each
Course Topic:
User Maintenance and Troubleshooting
Price (USD)
130.00
Each

The Dionex™ AS-AP autosampler eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The Dionex™ AS-AP autosampler basic operations and user maintenance with Chromeleon™ eLearning course provides new users with an overview of features and important guidelines of operation of the Thermo Scientific™ Dionex™ AS-AP Autosampler. Topics discussed in this course include:

  • Overview of main features
  • Description of main operating principles (Simultaneous vs. sequential injection mode, Push vs. Pull injection mode, Descriptions of its multiple optional features)
  • Instrument control in Chromeleon™ 7 CDS
  • Basic operational guidelines

The Dionex™ AS-AP Autosampler user maintenance and troubleshooting eLearning course provides new users with tips and video demonstrations on routine user maintenance procedures for a Thermo Scientific™ Dionex™ AS-AP Autosampler configured in Push or Pull mode. Topics covered in this presentation:

  • Basic overview of the sampling principle
  • Common replacement parts
  • General checks and routine maintenance
  • Replacement part procedures
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicUser Maintenance and Troubleshooting
DescriptionDionex™ ASAP™ Autosampler user maintenance and troubleshooting
FormatSelf-paced eLEarning
LanguagesEnglish
TechniqueIon Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelRoutine
Duration<30 min.
For Use With (Equipment)Dionex Aquion ICS
Product LineThermo Scientific
Product TypeChromatography
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.