Dionex™ AutoTrac™ 280 PFAS eLearning courses
Thermo Scientific™

Dionex™ AutoTrac™ 280 PFAS eLearning courses

The Dionex™ AutoTrace™ 280 PFAS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Catalog NumberCourse Topic
TAT28PBOPBasic Operations
Catalog number TAT28PBOP
Price (USD)
130.00
Each
Course Topic:
Basic Operations
Price (USD)
130.00
Each

The Dionex™ AutoTrace™ 280 PFAS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The Dionex™ AutoTrace™ 280 PFAS Basic Operation eLearning course includes a series of video demonstrations to provide the new user with a basic introduction to Solid-Phase Extraction (SPE) and the Dionex™ AutoTrace™ 280 instrument. Upon completion of this presentation, the user will have a comprehensive understanding of:

  • What is SPE and how it is used
  • The 4 steps of solid-phase extraction
  • The flow path of the AutoTrace™ 280 instrument
  • How to operate the software to run a sample

This Basic Operations course is similar to the Dionex AutoTrace™ 280 Basic Operations.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionDionex AutoTrace 280 PFAS basic operations
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueSolid Phase Extraction
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration2 to 4 hr.
For Use With (Equipment)Dionex AutoTrace 280 PFAS
Product LineThermo Scientific
Product TypeSample Preparation
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.