TDE Series Ultra-Low Temperature Freezer Package with Racks and Boxes - FAQs
TDE Series Ultra-Low Temperature Freezer Package with Racks and Boxes - FAQs
View additional product information for TDE Series Ultra-Low Temperature Freezer Package with Racks and Boxes - FAQs (TDE30086LDRK, TDE30086LARK, TDE30086FARK, TDE30086FDRK)