TDE Series -40°C Ultra-Low Temperature Freezer, 549 L, 230 V - FAQs
View additional product information for TDE Series -40°C Ultra-Low Temperature Freezer - FAQs (TDE40040LA, TDE40040LD, TDE40040LV, TDE40040FA, TDE40040FD, TDE40040FV)