TDE Series Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs
TDE Series Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs
View additional product information for TDE Series Ultra-Low Temperature Freezer Package with Racks, Boxes, and LN2 Back-up System - FAQs (TDE50086FARLN)