TDE Series -86°C Ultra-Low Temperature Chest Freezers - FAQs
View additional product information for TDE Series -86°C Ultra-Low Temperature Chest Freezers, General Purpose - FAQs (TDEC39686FV, TDEC39686FA, TDEC06386FV, TDEC25286FA, TDEC39686FD, TDEC25286FD, TDEC06386FA, TDEC25286FV)