EXTREVA™ ASE™ eLearning course
Thermo Scientific™

EXTREVA™ ASE™ eLearning course

EXTREVA™ ASE™ eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Catalog NumberDuration
TEASEBOP<30 min.
Catalog number TEASEBOP
Price (USD)
260.00
Each
Duration:
<30 min.
Price (USD)
260.00
Each

EXTREVA™ ASE™ eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The EXTREVA™ ASE™ Basic Operations eLearning course
Provides an introduction to the Thermo Scientific™ EXTREVA™ ASE™ Accelerated Solvent Extractor and how it works. The basic components of the system will be explained. How the Extreva ASE works and how it is different from the ASE-350 will be described.

  • Overview of the Extreva ASE and its components
  • How the Extreva ASE evaporation works
  • How the Extreva ASE is different from the ASE-350
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionEXTREVA ASE basic operations
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueAccelerated Solvent Extraction
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration<30 min.
For Use With (Equipment)EXTREVA ASE
Product LineThermo Scientific
Product TypeSample Preparation
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not have a thermofisher.com account?

If the user you have assigned administrative rights to does not have a thermofisher.com account, they will have to register for an account on thermofisher.com to assign the eLearning course. This account does not need to be a verified account and can be created by entering the following information:

- First Name
- Last Name
- Email address
- Password

The email address used to create the account will need to be the same email used to delegate the administration of the eLearning course.