Extreva™ ASE™ User Maintenance and Troubleshooting eLearning Course
Thermo Scientific™

Extreva™ ASE™ User Maintenance and Troubleshooting eLearning Course

The eLearning courses for the Thermo Scientific™ EXTREVA™ ASE™ Accelerated Solvent Extractor provide foundational knowledge and basic skills you need to use and benefit from your instrument.
Have Questions?
Catalog NumberCourse TopicCourse Level
TEASEMTSUser Maintenance and TroubleshootingRoutine
Catalog number TEASEMTS
Price (USD)
800.00
Each
Course Topic:
User Maintenance and Troubleshooting
Course Level:
Routine
Price (USD)
800.00
Each

The eLearning courses for the Thermo Scientific™ EXTREVA™ ASE™ Accelerated Solvent Extractor provide foundational knowledge and basic skills you need to use and benefit from your instrument. You will find a series of eLearning courses that can cover different topics such as foundational theory, basic operations principles, and/or user maintenance and troubleshooting.

The Thermo Scientific™ EXTREVA™ ASE™ Accelerated Solvent Extractor Maintenance and Troubleshooting eLearning course
Provides instruction and video demonstrations on recommended routine maintenance procedures for the Thermo Scientific™ EXTREVA™ ASE™ instrument

Topics covered in this presentation include:

  • Cell maintenance
  • Check valve replacement
  • Seal replacement
  • Backpressure valve rebuild
  • Electronic rotary valve (ERV) maintenance
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicUser Maintenance and Troubleshooting
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration30 to 60 min.
For Use With (Equipment)EXTREVA ASE
Product LineThermo Scientific
Product TypeSample Preparation
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach