Dionex™ Easion™ / ICS-900 eLearning Courses - FAQs
Dionex™ Easion™ / ICS-900 eLearning Courses - FAQs
View additional product information for Dionex™ Easion™ and ICS-900 eLearning Courses - FAQs (TEASFTH, TEASBOP, TEASMTS)
1 product FAQs found
How do I purchase eLearning courses online with a purchase order (PO)?
To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.