Orbitrap™ Eclipse Tribrid Mass Spectrometer eLearing Courses
Thermo Scientific™

Orbitrap™ Eclipse Tribrid Mass Spectrometer eLearing Courses

The Orbitrap Eclipse™ Tribrid™ Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principles and/or user maintenance and troubleshooting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicCourse Level
TECLBOPBasic OperationsRoutine
TECLFTHFoundational TheoryFoundation
Catalog number TECLBOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Course Level:
Routine
Price (USD)
490.00
Each

The Orbitrap Eclipse™ Tribrid™ Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.

The Orbitrap™ Eclipse Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument, which combines industry-leading versatility and usability with the exceptional performance afforded by its built-in intelligence and revolutionary Thermo Scientific™ Tribrid™ architecture to deliver an instrument designed to address the most difficult analytical challenges. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principles and/or user maintenance and troubleshooting.

Specifications
Access Availablity30 days upon end user first access to the course
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration60 to 120 min.
For Use With (Equipment)Orbitrap Eclipse Tribrid Mass Spectrometer
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach