EASY-nLC™ 1200 eLearning courses
Thermo Scientific™

EASY-nLC™ 1200 eLearning courses

The EASY-nLC™ 1200 eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TEZLCBOPBasic Operations
TEZLCMTSUser Maintenance and Troubleshooting
TEZLCFTHFoundational Theory
Catalog number TEZLCBOP
Price (USD)
300.00
Each
Course Topic:
Basic Operations
Price (USD)
300.00
Each

The EASY-nLC™ 1200 e eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The EASY-nLC™ 1200 Foundational Theory eLearning course provides an introduction to the basics of chromatography with a focus on liquid chromatography. This course covers the basic principles of chromatography, an introduction to High Pressure Liquid Chromatography (HPLC) theory and nano HPLC theory as well as a discussion on factors that influence liquid chromatography separation. Some of the topics covered in this eLearning course include:

  • Identifying key components of a chromatogram and the components that cause variations
  • Different separation types based on types of columns, packing materials and flow rates
  • How nano HPLC works and where it is used
  • Basic examples of separation and the influence of altering chromatography conditions

This Foundational Theory eLearning course is similar to the Vanquish™ Neo UHPLC System and UltiMate™ 3000 RSLCnano Foundational Theory courses.

This Foundational Theory eLearning course is similar and applicable to all nano HPLC instruments; e.g. UltiMate™ 3000 RSLCnano, and Vanquish™ Neo UHPLC.

The EASY-nLC™ 1200 Basic Operations eLearning course introduces the new user the to the Thermo Scientific™ EASY-nLC™ 1200 System hardware & software. It covers the process of creating a robust method and understanding the recommended parameters. This course also covers the tubing, columns, and connections used. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. Some of the topics include:

  • Method Run, and its settings
  • Importance of each setting and the implications of non-compliance with the rules
  • Tubing & columns used, including nanoViper
  • Familiarization with The Complete and Easy Guide to Configuring Your Thermo Scientific™ NanoLC for Mass Spectrometric Analysis
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionEASY-nLC 1200 basic operation
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueLiquid Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration30 to 60 min.
For Use With (Equipment)EASY-nLC 1200
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.