iConnect™ Detectors for TRACE™ 1300 and 1600 Series GC eLearning courses
Thermo Scientific™

iConnect™ Detectors for TRACE™ 1300 and 1600 Series GC eLearning courses

iConnect™ Detectors for TRACE™ 1300 and 1600 Series GC eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
TGCTCDMTSIConnect Thermal Conductivity Detector
TGCECDMTSIConnect Electron Capture Detector
TGCFIDMTSIConnect Flame Ionization Detector
Catalog number TGCTCDMTS
Price (USD)
260.00
Each
For Use With (Equipment):
IConnect Thermal Conductivity Detector
Price (USD)
260.00
Each

iConnect™ Detectors for TRACE™ 1300 and 1600 Series GC eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The iConnect™ Flame Ionization Detector for TRACE™ 1300 and 1600 Series GC basic operations and user maintenance eLearning course provides demonstrations of the installation, maintenance, and troubleshooting of the iConnect™ Flame Ionization Detector (FID) for TRACE™ 1300 and 1600 Series GC systems. Topics covered in the course include:

  • Familiarization of detector properties
  • Switching to the FID detector
  • Maintenance tasks: component replacement, flow monitoring, checkout

The iConnect™ Thermal Conductivity Detector for TRACE™ 1300 and 1600 Series GC basic operations and user maintenance eLearning course provides demonstrations of the installation, and tips on the routine operation and troubleshooting of the iConnect™ Thermal Conductivity Detector (TCD) for TRACE™ 1300 and 1600 Series GC systems. Topics covered in this course include:

  • Familiarization with detector properties
  • Switching to the TCD detector
  • Routine use: filament care, checkout, and flow monitoring

The iConnect™ Electron Capture Detector for TRACE™ 1300 and 1600 Series GC basic operations and user maintenance eLearning course provides a user demonstration of the installation, maintenance, and troubleshooting of the iConnect™ Electron Capture Detector (ECD) for TRACE™ 1300 and 1600 Series GC systems. Topics covered in this course include:

  • Familiarization with detector properties
  • Switching to the FID detector
  • Maintenance tasks: component replacement, flow monitoring, checkout
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionIConnect Thermal Conductivity Detector for TRACE 1300 and 1600 Series GC basic operations and user maintenance
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueGas Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelRoutine
Duration<30 min.
For Use With (Equipment)IConnect Thermal Conductivity Detector
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.