Gallery™ and Gallery Plus™ Discrete Analyzers eLearning Courses
Thermo Scientific™

Gallery™ and Gallery Plus™ Discrete Analyzers eLearning Courses

The Thermo Scientific™ Gallery™ and Gallery Plus™ Discrete Analyzers eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicCourse LevelDuration
TGDIAFTHFoundational TheoryFoundation30 to 60 min.
TGDIABOPBasic OperationsRoutine60 to 120 min.
Catalog number TGDIAFTH
Price (USD)
260.00
Each
Course Topic:
Foundational Theory
Course Level:
Foundation
Duration:
30 to 60 min.
Price (USD)
260.00
Each

The Thermo Scientific™ Gallery™ and Gallery Plus™ Discrete Analyzers eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.

The Thermo Scientific™ Discrete Analyzers Gallery™ and Gallery Plus™ eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations.

The Discrete Analyzers: Gallery™ and Gallery Plus™ Foundational Theory eLearning course
Introduces the basic principles of the Gallery instruments and how they utilize discrete photometric and electrochemical methods for automated wet chemistry analysis. Some of the topics covered in this eLearning course include:

  • Understanding the analytical technique Gallery instruments use to measure samples
  • Differences between the Gallery instruments
  • How Gallery instruments work and where it is used
  • Understanding analytical capabilities with the Gallery instruments

The Discrete Analyzers: Gallery™ and Gallery Plus™ Basic Operations eLearning course
Introduces the new user the to the Gallery™ and Gallery Plus™ System. This course is designed for new users who want to learn how to use/run basic the instrument. Some of the topics include:

  • How to perform a basic calibration of an assay
  • Running an unknown sample against the calibration
  • Reporting results
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicFoundational Theory
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelFoundation
Duration30 to 60 min.
For Use With (Equipment)Gallery Discrete Analyzer
Product LineThermo Scientific
Product TypeChromatography
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach