iCAP™ 7000 Series ICP-OES eLearning Courses
Thermo Scientific™

iCAP™ 7000 Series ICP-OES eLearning Courses

The iCAP™ 7000 Series ICP-OES eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TICAP7BOPBasic Operations
TICAP7FTHFoundational Theory
TICAP7MTSUser Maintenance and Troubleshooting
Catalog number TICAP7BOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Price (USD)
490.00
Each

These eLearning courses are an introduction to the basic principles of ICP-OES and how the iCAP 7000 Series instruments use this technology.

The iCAP™ 7000 Series Basic Operations eLearning course
will provide you with the basic training on the theory, operation, and general maintenance of your instrument. Included in the e-Learning course are the following e-learning modules:

iCAP™ 7000+ ICP-OES:

  • Peristaltic Pump and Pump Tubing Maintenance
  • Concentric Glass Nebulizer
  • Glass Cyclonic Spray Chamber
  • Plasma Torch Assembly
  • Purged Optical Path (POP) Window Maintenance
  • ThermoFlex 900 Recirculating Chiller
  • Thermo Scientific™ Qtegra™ ISDS Software: Introduction to the Qtegra Software
  • Thermo Scientific™ Qtegra™ ISDS Software: Creating a LabBook
  • Thermo Scientific™ Qtegra™ ISDS Software: Reviewing and Exporting Data and Reports
  • iCAP 7000+ ICP-OES: Creating Calibration Standards Volumetrically
  • iCAP 7000+ ICP-OES: Startup and Shutdown Procedures
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
TechniqueInductively Couple Plasma – Optical Emission Spectroscopy
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration4 to 8 hr.
For Use With (Equipment)iCAP 7000 Series ICP-OES
Product LineThermo Scientific
Product TypeAtomic Spectroscopy
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.