iCE ™ 3400 AAS Atomic Absorption Spectrometer eLearning Courses
Thermo Scientific™

iCE ™ 3400 AAS Atomic Absorption Spectrometer eLearning Courses

The iCE™ 3400 AAS Atomic Absorption Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Catalog NumberCourse Topic
TICE34BOPBasic Operations
Catalog number TICE34BOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Price (USD)
490.00
Each

The iCE™ 3400 AAS Atomic Absorption Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The iCE™ 3400 AAS Atomic Absorption Spectrometer basic operations eLearning course will provide a familiarization of the instrument and its operation. It will also provide a more in-depth discussion of Atomic Absorption Theory, the components of the instrument, alignment and optimization and the spectrometer settings. Some of the topics included in this course include:

  • Basic Familiarization for the iCE 3000 series of instruments
  • Basic Familiarization for the iCE 3400
  • Atomic Absorption Theory
  • Alignment and Optimization
  • Spectrometer Settings
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptioniCE™ 3400 AAS Atomic Absorption Spectrometer basic operations
FormatSelf-paced eLearning
LanguagesEnglish
TechniqueAtomic Absorption Spectroscopy
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)iCE 3300 AAS
Product LineThermo Scientific
Product TypeAtomic Spectroscopy
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.