Dionex™ ICS-5000+ HPIC™ eLearning courses
Dionex™ ICS-5000+ HPIC™ eLearning courses
Dionex™ ICS-5000+ HPIC™ eLearning courses
Dionex™ ICS-5000+ HPIC™ eLearning courses
Thermo Scientific™

Dionex™ ICS-5000+ HPIC™ eLearning courses

Dionex™ ICS-5000+ HPIC™ eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicDuration
TICS5BOPBasic Operations30 to 60 min.
TICS5MTSUser Maintenance and Troubleshooting2 to 4 hr.
TICS5EBOPBasic Operations60 to 120 min.
TICS5FTHFoundational Theory30 to 60 min.
Catalog number TICS5BOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Duration:
30 to 60 min.
Price (USD)
490.00
Each

Dionex™ ICS-5000+ HPIC™ eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The Dionex™ ICS-5000+ HPIC™ Foundational Theory eLearning course is an introduction to the general principles of ion chromatography. This course provides a review of the basic principles of chromatography and the history of the development of ion chromatography (IC) technology. The standard components of the IC system and how these facilitate the successful separation of analytes will be covered. Additional concepts discussed include:

  • What is ion chromatography?
  • A detailed description of the ion exchange process
  • Effects of mobile & stationary phases
  • Basic overview of Reagent-Free™ Ion Chromatography (RFIC™) technology
  • IC columns and their factors affecting peak resolution
  • Principles of suppressed conductivity detection

This Foundational Theory eLearning course is similar to the the Dionex™ ICS-6000 HPIC™, the Dionex™ Integrion™ HPIC™, the Dionex™ Aquion™ ICS and the Dionex™ ICS-2100.

The Dionex™ ICS-5000+ HPIC™ basic operations conductivity detector with Chromeleon™ eLearning course will provide basic operational guidelines to run samples using a Thermo Scientific™ Dionex™ ICS-5000+ HPIC system with conductivity detection and an AS-AP sampler. This course will also provide a detailed step-by-step instruction checklist to install a new DRS 600 suppressor on your ion chromatography system. This course provides an overview, installation, operation, and troubleshooting guidelines for Reagent-Free™ Ion Chromatography (RFIC™) consumables often used in standard IC systems. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. Some of the topics covered include:

  • System startup: eluent preparation, pump priming, and system equilibration
  • Creating sequences & methods
  • Routine operation of RFIC™ devices (EGC, CR-TC, Suppressors)

Dionex™ ICS-5000+ HPIC™ basic operations electrochemical detector with Chromeleon™ eLearning course will provide basic operational guidelines to run samples using a Thermo Scientific™ Dionex™ ICS-5000+ HPIC™ system with electrochemical (amperometry) detection and a Dionex™ AS-AP autosampler. This course provides an overview, installation, operation, and troubleshooting guidelines for manually prepared eluents and the electrochemical detector cell. This course is designed for new users who want to learn how to use/run basic experiments on the instrument. Some of the topics covered include:

  • System startup: eluent preparation, pump priming, and system equilibration
  • Creating sequences & methods
  • Installation and setup of the ED cell and its components

Dionex™ ICS-5000+ HPIC™ user maintenance and troubleshooting eLearning course discusses fundamental troubleshooting principles and definitions. It provides a review of an overall description of an Ion Chromatography System setup with conductivity detection and discusses its vital signs. It also provides users with a compilation of routine maintenance and troubleshooting guidelines for ion chromatography systems with electrochemical detection (ED). Some of the topics covered include:

  • Troubleshooting tips that will help quickly resume operation to generate accurate, reliable data
  • Requirements for ED operation
  • Routine maintenance guidelines
  • Troubleshooting tips on most-probable symptoms/problems.

This User Maintenance and Troubleshooting course is similar to the Dionex™ Integrion™ HPIC™ and Dionex™ ICS-6000 HPIC™ User Maintenance and Troubleshooting courses.

Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
DescriptionDionex ICS-5000+ HPIC conductivity detector basic operations with Chromeleon
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueIon Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration30 to 60 min.
For Use With (Equipment)Dionex ICS-5000+ HPIC
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.