Forma™ Direct Heat CO<sub>2</sub> Incubator eLearning Course
Thermo Scientific™

Forma™ Direct Heat CO2 Incubator eLearning Course

Flexible, self-paced, effective option to train and certify your operators on Thermo Fisher equipment. These courses are designed and executed by certified and qualified expert trainers; covers basic installation, startup, operation and user maintenance.
Have Questions?
Catalog NumberDuration
TINCCO2FOF30 to 60 min.
Catalog number TINCCO2FOF
Price (USD)
-
Duration:
30 to 60 min.
Thermo Scientific™ Forma Direct Heat CO2 Incubators eLearning course provides you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning modules which cover different topics such as introduction and/or basic operations and/or user maintenance.

Thermo Scientific™ Forma Direct Heat CO2 Incubator eLearning course is designed to help you get the most out of your technology, no matter your skill level. Included in this course:

Getting Started

 

  • Basic setup and configuration
  • Navigating your device
  • Connecting the CO2 gas supply

 

Core Features

 

  • Utilizing key functions
  • Customizing settings
  • Performing calibration on the unit
  • Responding to alarms

 

User Maintenance

  • Cleaning the unit
  • Maintaining your device

 

eLearning Courses

Empowering You with Knowledge. Begin your journey to becoming a device expert! Utilize course certificates as a part of your compliance.

Specifications
Access Availablity1 year upon end-user first access to the course
Course TopicThermo Scientific™ Forma Direct Heat CO2 Incubators Foundation eLearning course
FormatSelf-paced learning
LanguagesEnglish
Time and DateDigital access through Online Learning Management System
Course LevelFoundation
Duration30 to 60 min.
For Use With (Equipment)Forma Direct Heat CO2 Incubators
Product LineThermo Scientific
Product TypeTraining
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach