ISQ™ EM Single Quadrupole MS eLearning Courses
Thermo Scientific™

ISQ™ EM Single Quadrupole MS eLearning Courses

ISQ™ EM Single Quadrupole MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TISLEMBOPBasic Operations
TISLEMFTHFoundational Theory
TISLEMMTSUser Maintenance and Troubleshooting
Catalog number TISLEMBOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Price (USD)
490.00
Each

ISQ™ EM Single Quadrupole MS eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The ISQ™ EM Single Quadrupole MS foundational theory eLearning course introduces the ISQ™ EM and the basics of mass spectrometry. This course provides an introduction to the ISQ™ EM and the Ion Chromatography and Liquid Chromatography instruments which can be used with it. Included in this course:

  • Definition of mass spectrometry
  • The four phases of mass spectrometry and discuss the role of each of them in the process.

This Foundational Theory course is similar to the ISQ™ EC Single Quadrupole MS Foundational Theory course.

The ISQ™ EM Single Quadrupole MS basic operations with Chromeleon™ eLearning course will provide an overview of the instrument, method considerations and method creation. This course will also cover basic data processing in Chromeleon. Some of the topics included in the course include:

  • An introduction to the different components of the ISQ Mass Spectrometer
  • LC-MS Method Considerations
  • Chromeleon Control and Method Creation
  • Data Processing in Chromeleon.

The topics covered in this course will be similar to those covered in the ISQ™ EC Single Quadrupole MS basic operations with Chromeleon course.

The ISQ™ EM Single Quadrupole MS user maintenance and troubleshooting eLearning course provides basic maintenance and troubleshooting guidelines for Thermo Scientific™ ISQ™ EM Single Quadrupole MS.

Topics covered in this course include:

  • Routine and preventative maintenance tasks:
  • ESI probe capillary replacement
  • APCI probe capillary replacement
  • APCI needle replacement
  • System shutdown
  • Front optics removal, disassembly, cleaning, and reassembly
  • Routine diagnostics and troubleshooting guidelines:
  • General troubleshooting information
  • AutoTune diagnostics
  • Troubleshooting gas leaks
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
TechniqueLiquid Chromatography Mass Spectrometry
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration60 to 120 min.
For Use With (Equipment)ISQ EM Single Quadrupole MS
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not have a thermofisher.com account?

If the user you have assigned administrative rights to does not have a thermofisher.com account, they will have to register for an account on thermofisher.com to assign the eLearning course. This account does not need to be a verified account and can be created by entering the following information:

- First Name
- Last Name
- Email address
- Password

The email address used to create the account will need to be the same email used to delegate the administration of the eLearning course.