UV-Visible Detectors eLearning Courses
Thermo Scientific™

UV-Visible Detectors eLearning Courses

UV-Visible Detectors eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse TopicFor Use With (Equipment)
TLCVWDTHFoundational TheoryVariable Wavelength Detectors
TLCDADBOPBasic OperationsDiode Array Detectors
TLCDADFTHFoundational TheoryDiode Array Detectors
TLCVWDBOPBasic OperationsVariable Wavelength Detectors
Catalog number TLCVWDTH
Price (USD)
75.00
Each
Course Topic:
Foundational Theory
For Use With (Equipment):
Variable Wavelength Detectors
Price (USD)
75.00
Each

UV-Visible Detectors eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.

The Variable Wavelength Detectors Foundational Theory eLearning course provides an overview of the theory behind the operation of variable wavelength detectors. Included in this course is:

  • Fundamental theory and purpose of a VWD
  • The optical design used by VWD and how a VWD works in principle
  • Critical features of the VWD including working range and data acquisition modes VWDs can use

The Variable Wavelength Detectors basic operations and best practices eLearning course will provide insight into programming the detector for operation. It also includes critical items necessary to ensure optimized data quality generated by the Variable Wavelength Detector (VWD) and special considerations to which apply to Variable Wavelength Detectors. Some of the topics covered in this course include:

  • Key parameters required to directly control the detector
  • Appropriate settings for automated analysis
  • Concepts of data collection rate and response time
  • General troubleshooting
  • Factors that contribute to symptoms such as noise, drift and signal intensity
  • The process for cleaning VWD flow cells

The Diode Array Detectors Foundational eLearning course is an introduction to DAD operation, design and features. Included in this course is:

  • Fundamental theory and purpose of a DAD
  • Optical design used by DAD and how a DAD works in principle
  • Critical features of the DAD including working range and data acquisition modes DADs can use

The Diode Array Detectors Basic Operations and best practices eLearning course will provide details on programming the detector for operation. It also includes critical items to optimize data quality and special considerations when using a Diode Array Detector. Some of the modules in the course include:

  • Programming appropriate settings for automated analysis
  • Concepts of data collection rate and response time
  • Important solvent properties when working with UV/Vis detectors
  • Principles affecting the extent of photo-oxidation of compounds
  • Critical operating parameters such as eluent temperature and back pressure when using light pipe flow cells
Specifications
Access Availablity30 days upon end-user first access to the course
Course TopicFoundational Theory
DescriptionDiode Array Detectors foundational theory
FormatOnline Self-paced eLearning
LanguagesEnglish
TechniqueLiquid Chromatography
Time and DateOn-demand upon course end-user registration confirmation
Course LevelFoundation
Duration<30 min.
For Use With (Equipment)Variable Wavelength Detectors
Product LineThermo Scientific
Product TypeChromatography
Target AudienceNew user seeking training on instrument operation and basic experimentation
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I purchase eLearning courses online with a purchase order (PO)?

To purchase an eLearning course with a PO, you need to have a purchasing account on thermofisher.com. For detailed steps on how to register and set up a purchasing account, please review this resource page (https://www.thermofisher.com/us/en/home/support/how-to-register-for-account.html). After your purchasing account has been set up, follow the instructions in this How-To-Guide (https://www.thermofisher.com/us/en/home/products-and-services/services/unity-lab-services/equipment-instrument-education/elearning-how-to-guide.html) for finding and ordering eLearning courses online with a PO.

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.