You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Western Blot Products
▸
Western Blot and Nucleic Acid Membranes and Filter Paper
▸
Owl™ Transfer Membranes
Actual product may vary
Thermo Scientific™
Owl™ Transfer Membranes
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
Have Questions?
Change view
Catalog Number
Packaging Type
TM151-1
Roll
TM151-4
Sheets (15 x 15cm)
TM151-5
Sheets (10 x 10cm)
TM
Transfer Membrane; Complete system
4 Options
Catalog number
TM151-1
Price (USD)
544.50
Online exclusive
605.00
Save 60.50
Pack of
Add to cart
Packaging Type:
Roll
Price (USD)
544.50
Online exclusive
605.00
Save 60.50
Pack of
Add to cart
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™.
This membrane provides the highest signal-to-noise ratio, which allows the use of mild blocking procedures.
Immobilon-P has high protein retention, is compatible with all standard visualization methods and can be used for protein sequencing.
Immobilon is available in minigel and large-format gel sizes.
Specifications
Description
26.5cm x 3.75m; Roll
For Use With (Application)
Western blotting
Packaging Type
Roll
Dimensions (LxW)
3.75 m x 26.5 cm
Format
Roll
Length (Metric)
26.5 cm
Material
Immobilon-P, PVDF
Product Line
Owl
Width (Metric)
3.75 m
Unit Size
Pack of