Orbitrap™ Exploris™ GC Mass Spectrometer eLearning Courses
Thermo Scientific™

Orbitrap™ Exploris™ GC Mass Spectrometer eLearning Courses

The Thermo Scientific™ Orbitrap™ Exploris™ GC Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument. You will find a series of eLearning courses which can cover different topics such as foundational theory and/or basic operations principle and/or user maintenance and troubleshooting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCourse Topic
TOEXGBOPBasic Operations
TOEXGFTHFoundational Theory
TOEXGMTSUser Maintenance and Troubleshooting
Catalog number TOEXGBOP
Price (USD)
490.00
Each
Course Topic:
Basic Operations
Price (USD)
490.00
Each

The Thermo Scientific™ Orbitrap™ Exploris™ GC Mass Spectrometer eLearning courses provide you with foundational knowledge and basic skills that you need to use and benefit from your instrument.

The Orbitrap™ Exploris™ GC Mass Spectrometer Basic Operations course
Provides you with the basic guidelines for the routine operation of the Thermo Scientific™ Orbitrap™ Exploris™ GC Mass Spectrometer. Upon completion of this course, you will understand:

  • How to power ON the instrument.
  • The process of bakeout and vacuum check.
  • Instrument controls and the software user interface.
  • How to perform tuning and mass calibration.
  • How to create an instrument method using Chromeleon CDS.

The Orbitrap™ Exploris™ GC Mass Spectrometer foundational theory eLearning course
Offers detailed information on the theory and fundamental principles of the Thermo Scientific™ Orbitrap™ Exploris™ GC Mass Spectrometer, including:

  • A comprehensive overview of the timeline and history of Orbitrap technology and its adaptation for Gas Chromatography applications.
  • An introduction to the platform and an overview of the ion pathway, specifically for the Orbitrap GC Exploris.

Fundamental principles of the instrument, focusing on four key aspects:

  • Ion production (Electron Ionization or Chemical Ionization)
  • Transportation of the ions from their formation to the rest of the analyzer
  • Selection and filtration of masses in the quadrupole, along with an examination of the HCD cell and its significance in facilitating MS/MS experiments
  • Functionality of the C-trap and the Orbitrap, and how they work harmoniously to ensure the detection of selected ions.

The Thermo Scientific™ Orbitrap™ Exploris™ GC MS Maintenance and Troubleshooting eLearning course
Provides detailed instructions on basic maintenance procedures and troubleshooting guidelines presented in three sections:

  • The necessary steps to safely shut down the Orbitrap Exploris GC Mass Spectrometer
  • How to perform basic maintenance and troubleshooting tasks
  • How to perform tuning and mass calibration
Specifications
Access Availablity30 days upon end user first access to the course
Course TopicBasic Operations
FormatSelf-paced eLearning
LanguagesEnglish
Course LevelRoutine
Duration60 to 120 min.
For Use With (Equipment)Orbitrap Exploris 240 GC-MS
Product LineThermo Scientific
Product TypeMass Spectrometry
Target AudienceIntermediate user seeking to benefit from good instrument performance
Training LocationDigital access through Online Learning Management System
Unit SizeEach

Frequently asked questions (FAQs)

How do I change to whom an eLearning course for analytical instruments and lab equipment is assigned?

Access Credit Manager to manage the eLearning course assignment. Select the eLearning course order and click on the Assign credits button to remove the existing user assignment.
Note: You can only change the assigned user if they have not yet accessed the eLearning course.

Can I assign an eLearning course for analytical instruments and lab equipment to more than one person?

You can assign an eLearning course for analytical instruments and lab equipment to more than one person only if you purchased multiple quantities. The number of quantities purchased equals the number of users that can be assigned to the course.

How long do I have to assign a purchased eLearning course for analytical instruments and lab equipment to a user?

You have one year from the date of purchase to assign an eLearning course for analytical instruments and lab equipment to a user.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not receive the email on how to manage eLearnings?

Please verify the email address used for delegating. If you are unable to locate the email, you can access Credit Manager directly.

What if the user I am delegating an eLearning course for analytical instruments and lab equipment to does not have a thermofisher.com account?

If the user you have assigned administrative rights to does not have a thermofisher.com account, they will have to register for an account on thermofisher.com to assign the eLearning course. This account does not need to be a verified account and can be created by entering the following information:

- First Name
- Last Name
- Email address
- Password

The email address used to create the account will need to be the same email used to delegate the administration of the eLearning course.