Tokai Hit™ ThermoPlate™ System - FAQs
Tokai Hit™ ThermoPlate™ System - FAQs
View additional product information for Tokai Hit™ ThermoPlate™ System - FAQs (TPXEVOS)
4 product FAQs found
What objectives can I use with the Tokai Hit Thermoplate?
We recommend using long working distance (LWD) objectives as the glass heating plate is 0.5 mm thick, including the thickness of the culture plate or glass bottom dishes.
How can I clean the Tokai Hit Thermoplate and EVOS Stage Plate?
Wipe the thermoplate and vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol.
Can I place the EVOS Light Shield box over the Tokai Hit Thermoplate and stage holder?
Yes, you can place the EVOS Light Shield box over the Tokai Hit Thermoplate and stage holder.
Can I use the Tokai Hit Thermoplate and EVOS Stage Plate on the EVOS FL Auto Imaging System?
Yes you can, but we also offer the EVOS Onstage Incubator (Cat No. AMC1000) as a better option to control not just the temperature but also humidity and gas concentration for the EVOS FL Auto Imaging System. The EVOS Onstage Incubator cannot be used on the EVOS XL and FL imaging systems.