TruNarc™ Handheld Narcotics Analyzer - FAQs
TruNarc™ Handheld Narcotics Analyzer - FAQs
View additional product information for TruNarc™ Handheld Narcotics Analyzer - FAQs (TRUNARC)
10 product FAQs found
What is Raman spectroscopy?
In Raman spectroscopy, an unknown sample of material is illuminated with monochromatic (single wavelength or single frequency) laser light, which can be absorbed, transmitted, reflected, or scattered by the sample. Light scattered from the sample is due to either elastic collisions of the light with the sample's molecules (Rayleigh scatter) or inelastic collisions (Raman scatter). Whereas Rayleigh scattered light has the same frequency (wavelength) of the incident laser light, Raman scattered light returns from the sample at different frequencies corresponding to the vibrational frequencies of the bonds of the molecules in the sample.
If you wish to learn more about Raman spectroscopy, visit our online Raman Spectroscopy Academy (https://www.thermofisher.com/us/en/home/industrial/spectroscopy-elemental-isotope-analysis/spectroscopy-elemental-isotope-analysis-learning-center/molecular-spectroscopy-information/raman-technology.html), where you will find basic Raman tutorials, advanced Raman webinars on sample applications, and a helpful instrument guide.
How long does the TruNarc Handheld Narcotics Analyzer battery last?
The TruNarc Handheld Narcotics Analyzer has a 3.7 V internal rechargeable battery pack that lasts up to 10 hours.
Can the TruNarc Handheld Narcotics Analyzer scan through packaging?
Yes. Because the TruNarc Handheld Narcotics Analyzer employs non-destructive, non-contact Raman technology, it can scan directly through sealed glass and plastic containers. This feature helps reduce exposure to ensure officer safety, and is particularly useful for preserving evidence.
Can the results obtained from the TruNarc Handheld Narcotics Analyzer be used in court?
Depending on jurisdiction, TruNarc Handheld Narcotics Analyze test results may be allowed at preliminary hearings.
The TruNarc analyzer captures all scan results, including time-and-date stamp and system selfchecks to help expedite prosecution. Once a substance is analyzed, full results are automatically stored for reporting and evidence submission. Rapid results combined with automated reports can streamline the path to prosecution, reducing administrative burden and dramatically impacting the time and expense of drug-related arrests.
What are the limits of the technology used in the TruNarc Handheld Narcotics Analyzer?
Raman spectroscopy is best used as a primary analysis technique for sealed containers, aqueous solutions, and white and light powders. The TruNarc Handheld Narcotics Analyzer also uses a Type H kit to analyze substances with high fluorescence.
What is the technology used in the TruNarc Handheld Narcotics Analyzer?
The TruNarc Handheld Narcotics Analyzer uses lab-proven Raman spectroscopy. Raman is a vibrational spectroscopy technique where a single wavelength laser is focused on a sample. The laser excites the bonds of a molecule, which generates measurable scattered light to identify the material in question. Raman is a highly effective method for reliable identification of an unknown substance based on its underlying chemistry.
Raman spectroscopy has been extensively deployed for use in homeland security applications, largely due to the high chemical specificity, with each chemical substance having a unique spectral fingerprint.
How long does it take to get results with the TruNarc Handheld Narcotics Analyzer?
The TruNarc Handheld Narcotics Analyzer often presents point and shoot test results in seconds although some samples could take up to a minute.
Can you update the TruNarc Handheld Narcotics Analyzer library of items?
In order to maintain chain of custody and ensure court admissible results, the TruNarc Handheld Narcotics Analyzer includes a closed loop library. Library updates are made on a regular basis to ensure users have access to the latest emerging threats.
How many items are in the TruNarc analyzer library?
The expanded v1.10 library adds field identification capability for many new high priority alarm substances including several fentanyl derivatives, carfentanil, acrylfentanyl, fentanyl precursors (NPP and ANPP) and pharmaceutical variants (Alfentanil and Sufentanil). With the v1.10 library, the TruNarc Handheld Narcotics Analyzer enables officers, customs, border control, and other personnel to scan more than 530 suspected controlled substances in a single, definitive test.
Can the TruNarc Handheld Narcotics Analyzer identify fentanyl and fentanyl variants?
Yes, the TruNarc Handheld Narcotics Analyzer can identify fentanyl and several fentanyl variants. The expanded v1.9 library adds field identification capability for many new high priority alarm substances including several fentanyl derivatives, carfentanil, acrylfentanyl, fentanyl precursors (NPP and ANPP) and pharmaceutical variants (Alfentanil and Sufentanil). With the v1.9 library, the TruNarc Handheld Narcotics Analyzer enables officers, customs, border control, and other personnel to scan more than 498 suspected controlled substances in a single, definitive test.