PTFE/Silicone Discs
PTFE/Silicone Discs
PTFE/Silicone Discs
Thermo Scientific™
PTFE/Silicone Discs
Combine the inertness of a PTFE coating with the resealability of silicone Thermo Scientific™ PTFE/Silicone Discs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Outer
TS-1272222 mm
TS-127088 mm
TS-1271212 mm
TS-1271313 mm
TS-1271616 mm
TS-1271818 mm
Catalog number TS-12722
Price (USD)
208.17
Pack of 72
Diameter (Metric) Outer:
22 mm
Price (USD)
208.17
Pack of 72
Ensure air-tight, inert seals with autoclavable Thermo Scientific™ PTFE/Silicone Discs. They are specifically designed to combine the resealability of silicone with the inertness of PTFE. These discs reseal instantly puncture after puncture, and the PTFE coating is structurally bonded so that no cement is leached or baked out of your sample after needle penetration. Many sizes are available to fit Thermo Scientific™ Reacti-Vial™ Small Reaction Vials, Thermo Scientific™ Screw Cap Septum Vials, and Thermo Scientific™ Hypo-ViaI™ Sample Storage Vials.
Additional Product Information
Each disc compresses to maintain a tight seal, forcing the PTFE coating to conform to the sealing surface.
Standard syringe and CC needles penetrate an entire disc with ease.
The PTFE layer is a full 10 mils thick to ensure protection
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
DescriptionDiameter: 22mm
For Use With (Equipment)25 and 40 mL Screw Cap Septum Vial, 240 mL (8 oz.) Screw Cap Bottle, 10 mL Reacti-Vial
Product TypeDisc
Thickness (Metric)10 mil
Diameter (Metric) Outer22 mm
MaterialPTFE, Silicone
Quantity72 Pk.
Unit SizePack of 72