Screw Cap Septum Vials
Thermo Scientific™

Screw Cap Septum Vials

For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
TS-130193.5 mL
TS-130287 mL
Catalog number TS-13019
Price (USD)
148.15
Pack of 72
Volume (Metric):
3.5 mL
Price (USD)
148.15
Pack of 72
For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials. Made of autoclavable borosilicate glass and available with a wide assortment of special closures and accessories, these vials are ideal for sample collection and small derivatization reactions for use with automated gas (GC) and liquid chromatography (LC), and for storage of reagents and standards under complete seal with instant syringe access. The heavy-duty, flip-top divider box provides easy access to vials, caps, and septa and offers a convenient sample storage center. Clear and amber vials are available.
Applications

The 40mL clear or amber vial with a Thermo Scientific Disc is suitable for discrete water sampling under U.S. EPA 40 CFR Parts 136 and 141.

The 1.5mL clear and amber vials fit GC and LC autosamplers that use standard 12 × 32 mm vials.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Closure Size13-425
Description3.5mL; Clear
Height (Metric)45 mm
Product TypeSeptum Vial
Volume (Metric) Total3.5 mL
Closure TypeScrew Cap
ColorClear
Diameter (Metric) Outer15 mm
MaterialBorosilicate Glass
Volume (Metric)3.5 mL
Unit SizePack of 72