Open-Top Screw Caps
Open-Top Screw Caps
Open-Top Screw Caps
Thermo Scientific™

Open-Top Screw Caps

Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
TS-132153.5 mL
TS-132167 mL
TS-132183 mL
TS-1321910 mL
Catalog number TS-13215
Price (USD)
61.31
Pack of 72
Volume (Metric):
3.5 mL
Price (USD)
61.31
Pack of 72

Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.

  • Fit into Reacti-Vial™ small reaction vials
  • Used with PTFE/Rubber laminated discs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure Size13 mm
DescriptionFits 0.3 and 1.0mL Reacti-Vials, 3.5mL Screw Cap Septum Vial
For Use With (Equipment)0.3 and 1 mL Reacti-Vials, 3.5 mL Screw Cap Septum Vials
Product TypeOpen Top Screw Cap
Volume (Metric)3.5 mL
Unit SizePack of 72